missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
23.80 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Lanthanum(III) bromide is used as a scintillation material in certain applications. It is used in gamma spectroscopy radioisotope detection. It is used as a source of lanthanum in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
Solubility Information | Soluble in water. |
IUPAC Name | lanthanum(3+);tribromide;hydrate |
Formula Weight | 378.62 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Assay | (REO) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'