missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody
LanthaScreen™ Tb-anti-phospho-p53 [pSer15] Antibody
4280.00 € - 20610.00 €
Specifications
Antigen | Ser/Thr kinases |
---|---|
Classification | Monoclonal |
Conjugate | Tb (Terbium) |
Regulatory Status | RUO, RUO |
Primary or Secondary | Primary |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10033904
|
Invitrogen
PV5128 |
25 μg |
4280.00 €
Each |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10083614
|
Invitrogen
PV5129 |
1 mg |
20610.00 €
Each |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The p53 tumor suppressor protein is an essential component of many different signaling pathways. The LanthaScreen™ antibody detects the phosphorylation of p53 at Ser15 by a range of kinases including DNA-PK, ATM, and ATR.For use in combination with the Fluorescein-p53 (Ser15) peptide substrate (PV5132) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Ser/Thr kinases | |
Tb (Terbium) | |
Primary | |
150kDa |
Monoclonal | |
RUO, RUO | |
–20°C | |
1 Each |
For Research Use Only. Not for use in diagnostic procedures.