missing translation for 'onlineSavingsMsg'
Learn More

LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit

LanthaScreen™ Tb-p4E-BP1 (pThr46) Antibody Kit

Brand:  Invitrogen PV4757

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 10053143

  • 4020.00 € / Each
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A part of the PI3K/AKT Pathway, 4E-BP1 (EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1) is a cytosolic protein that is phosphorylated at Thr46 by mTORC1 upon stimulation of the pathway.

For use in combination with the GFP-4E-BP1 physiological protein substrate (PV4759) to evaluate inhibitors in medium to high throughput screening applications. An optimized mTor assay buffer (PV4794) is also available.
Specifications

Specifications

Serine/threonine kinase
0.52mg/mL
150kDa
RUO, RUO
Primary
–20°C
Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay
Tb (Terbium)/GFP (EmGFP)/Lanthanide Chelates
25 μg
Target and Lead Identification and Validation
1 Kit(s)
384-well Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.