missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LanthaScreen™ Tb-peIF2a (pSer52) Antibody
LanthaScreen™ Tb-peIF2a (pSer52) Antibody
4290.00 € - 20500.00 €
Specifications
Antigen | Serine/threonine kinase |
---|---|
Concentration | 0.47mg/mL |
Packaging | 1 Each |
Primary or Secondary | Primary |
Content And Storage | Terbium-labeled peIF2a (pThr46) antibody in HEPES buffered saline. Store at -20°C. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10013574
|
Invitrogen
PV4815 |
25 μg |
4290.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023574
|
Invitrogen
PV4816 |
1 mg |
20500.00 €
Each |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis. In response to a cellular stress, phosphorylation of eIF2alpha at Ser52 occurs by the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2) to prohibit the initiation of protein synthesis.For use in combination with the GFP-EIF2alpha physiological protein substrate (PV4809) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Serine/threonine kinase | |
1 Each | |
Terbium-labeled peIF2a (pThr46) antibody in HEPES buffered saline. Store at -20°C. | |
Tb (Terbium)/Lanthanide Chelates | |
RUO, RUO |
0.47mg/mL | |
Primary | |
Kinase Assay | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.