Learn More
Thermo Scientific™ Large O-ring for Eutech™ ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Electrodes
Thermo Scientific™ offers Large O-ring replacement parts for ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Electrodes.
Brand: Thermo Scientific™ 32X246702
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Eutech™ Large O-ring is used for replacement parts in ECDOGEN-S and ECDOTPII-S Dissolved Oxygen Electrodes. These galvanic electrodes are designed to be rugged and long- lasting.
Features:
- Replacement Part for Dissolved Oxygen Process Electrodes
Specifications
ECDOGEN-S, ECDOTPII-S DO Electrode | |
Replacement Part |