missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Large Reverse Osmosis Filter Inserts
Protect large reverse osmosis systems from undissolved substances in water by using these large RO system filter insets.
130.00 €
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
15597259
|
Thermo Scientific™
50155512 |
Absolute Filter 0.5µm (10 in.) | Pre-filter Housings, 10 in. (06.5310, 06.5311, 06.5312) |
130.00 €
Each |
In Stock
Log in to see stock. |
||||
Description
Protect large reverse osmosis systems from undissolved substances in water by using Thermo Scientific™ large reverse osmosis filter insets.
Big Blue filters
- Big Blue pre-filters protect installations and systems from undissolved substances in water
- Contain special housing and typically are used in big flow rate CP systems (>1500L)
Ordering Information:
This product is not available in all regions
Specifications
Filter Insert, Large Reverse Osmosis | |
1500 L |