missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ LC PEEKsil™ Tubing

Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.

Brand:  Trajan Scientific and Medical™ 0624217

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268733

  • 97.60 € / Pack of 5


Estimated Shipment
17-08-2020
Add to basket

Description

Description

  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
Specifications

Specifications

5/Pk.
1.6 mm
200 mm
0.3 mm
Polymer-Sheathed Fused Silica
PEEKsil Tubing
Videos
Documents