Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class F1 with certificate
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Leaf Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™
Additional Details : Weight : 0.50000kg
Product Code. 15555109
  • 467.46 € / 1 unit
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Class F1 calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
F1
1 to 500 mg
1 mg to 500 mg