Learn More
Emma Safety Footwear LeMans XD Safety Shoes
Provides dry, fresh feet for up to 24 hours. Offers optimal ventilation, perspiration and moisture removal with Hydro-Tec™ Sanitized Silver. The LeMans is ideally suited to indoor working environments or for those working in Transport and Logistics.
Brand:  Emma Safety Footwear MM72756046
Additional Details : Weight : 1.66000kg
 View more versions of this product
Product Code. 15908931
  • 139.92 € / 1 pair
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • S1-Safety shoe for dry working conditions
  • GripForce™ Easy Twist outsole
  • Nubuck construction
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
Lemans XD
Oiled Nubuck
46