missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lincomycin hydrochloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
Lincomycin hydrochloride is used to study the inhibition of protein synthesis, bacterial antibiotic resistance and protein and surfactant binding. It has been used in the treatment of Staphylococcal, Streptococcal, and Bacterioides fragilis infections.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Lincomycin hydrochloride is a lincosamide compound with antibacterial activity originally identified in the actinomycete, Streptomyces lincolnensis, with activity against gram-positive cocci and anaerobic bacteria
Lincomycin inhibits bacterial protein synthesis by forming cross-links within the peptidyl transferase loop region of the 23S rRNA
Application
Lincomycin hydrochloride is commonly used to study the inhibition of protein synthesis, bacterial antibiotic resistance, and protein and surfactant binding
It has been used against Staphylococcal, Streptococcal, and Bacteroides fragilis species
It is protective against the development of mastitis in a mouse model of acute mastitis induced by S. aureus
Specifications
Type | Lincomycin Hydrochloride |
Physical Form | Crystalline Powder |
Beilstein | 4171650 |
Solubility Information | Soluble in water |
Formula Weight | 443 |
Color | White |
Quantity | 5 g |
Chemical Name or Material | Lincomycin hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'