missing translation for 'onlineSavingsMsg'
Learn More

Lithium cobalt(III) oxide, 97%, Thermo Scientific Chemicals

Catalog Number 11307667
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
12190-79-3
CoLiO2
97.871
MFCD00049786
BFZPBUKRYWOWDV-UHFFFAOYSA-N
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
23670860
lithium;oxido(oxo)cobalt
[Li+].[O-][Co]=O
This item is not returnable. View return policy
12190-79-3
CoLiO2
97.871
MFCD00049786
BFZPBUKRYWOWDV-UHFFFAOYSA-N
lithium colbaltite, lithium cobalt iii oxide, cobalt lithium oxide, cobaltate coo21-, lithium, cobalt lithium dioxide, coo2.li, lithium oxido oxo cobalt, lithium cobalt dioxide, lithotab cobaltoylolate
23670860
lithium;oxido(oxo)cobalt
[Li+].[O-][Co]=O

Lithium cobalt(III) oxide is a critical substance in the quest for high performance, advanced batteries. It is used as a lithium ion cathode material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue
Physical Form Powder
Quantity 250 g
Assay Percent Range 97%
Solubility Information Insoluble in water.
Formula Weight 97.88
Odor Odorless
Sensitivity Hygroscopic
Appearance Dark blue powder
Chemical Name or Material Lithium cobalt(III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'