missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 97%, for analysis, ACROS Organics™

Brand:  Acros Organics 218270025

Code : 29

Additional Details : CAS Number : 7789-24-4 Weight : 2.55000kg Transport : UN number : 3288 Chem class : 6.1 Pack group : III

Packaging Plastic bottle
Quantity 2.5kg

 View more versions of this product

Product Code. 11944171

  • 364.00 € / 2.50kg

Estimated Shipment: 05-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7789-24-4
25.93
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
lithium;fluoride
FLi
PQXKHYXIUOZZFA-UHFFFAOYSA-M
224478
[Li+].[F-]

Specifications

Lithium fluoride
96% min
LiF
15,5587
Solubility in water: 0.29g/100mL (20°C). Other solubilities: soluble in acids,insoluble in alcohol
[Li+].[F-]
25.93
25.93
97%
0.08% max.
0.07% max
2.6300g/mL
0.4% (200°C) (typical)
Plastic bottle
0.07% max.
Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol
1% max
1681.0°C
845.0°C
7789-24-4
FLi
01,933
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium;fluoride
224478
Powder
Analysis
1.0% max.
0.07% max.
0.03% max.
0.02% max.
0.04% max
0.4% max.
2.63
Trace analysis are typical values
White
2.5kg
Videos
SDS Documents
Certificates