missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog Number 11974151
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
1310-65-2
HLiO
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
3939
CHEBI:33979
[Li+].[OH-]
1310-65-2
HLiO
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
3939
CHEBI:33979
[Li+].[OH-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 462.0°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98%
UN Number 2680
Merck Index 15,559
Solubility Information Solubility in water: 113g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name lithium(1+) hydroxide
Formula Weight 23.95
Percent Purity 98%
Grade Pure
Packaging Plastic Bottle
Specific Gravity 1.43
Density 1.4300g/mL
Chemical Name or Material Lithium Hydroxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'