missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, 98%, pure, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3212801
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
1310-65-2
HLiO
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
3939
CHEBI:33979
[Li+].[OH-]
1310-65-2
HLiO
23.95
MFCD00011095
WMFOQBRAJBCJND-UHFFFAOYSA-M
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
3939
CHEBI:33979
[Li+].[OH-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 462.0°C
Color White to Yellow
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 98%
Merck Index 15,559
Solubility Information Solubility in water: 113g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name lithium;hydroxide
Formula Weight 23.95
Percent Purity 98%
Grade Pure
Packaging Plastic bottle
Specific Gravity 1.43
Density 1.4300g/mL
Chemical Name or Material Lithium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydroxide anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'