Learn More
Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH, ACROS Organics™
Brand: Acros Organics 199540050
Code : 29
Additional Details : CAS Number : 1310-66-3 Weight : 5.05000kg Transport : UN number : 2680 Chem class : 8 Pack group : II
Packaging | Plastic drum |
---|---|
Quantity | 5kg |
Chemical Identifiers
1310-66-3 | |
41.962 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
lithium;hydroxide;hydrate |
H3LiO2 | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
168937 | |
[Li+].O.[OH-] |
Specifications
Lithium hydroxide monohydrate | |
300ppm max. | |
LiOH·H2O | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
lithium;hydroxide;hydrate | |
168937 | |
Crystalline Powder | |
Extra Pure | |
500ppm max. | |
10ppm max. | |
3000ppm max. | |
500ppm max. | |
Cream to White or Yellow | |
5kg |
1310-66-3 | |
H3LiO2 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
[Li+].O.[OH-] | |
41.962 | |
41.95 | |
≥54% (LiOH) | |
3500ppm max. | |
0.1% max. | |
Plastic drum | |
Solubility in water: 109g/L (20°C). Other solubilities: 107g/L (0°C), 148g/L (100°C), slightly soluble in alcohol | |
920.0°C | |
462.0°C |
Safety and Handling
- Lithium hydroxide monohydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.