missing translation for 'onlineSavingsMsg'
Learn More

Lithium triethylborohydride, 1.7M solution in THF, Acros Organics

Brand:  Acros Organics 450698000

Code : NEW

Additional Details : CAS Number : 22560-16-3 Weight : 1.42000kg Transport : UN number : 3394 Chem class : 4.2 Pack group : I


Quantity 800mL

 View more versions of this product

Product Code. 15269305

  • 435.70 € / 800mL

Estimated Shipment
22-07-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

22560-16-3
C6H16BLi

Specifications

Lithium triethylborohydride
C6H16BLi
0.8900g/mL
Solubility in water: reacts.
800mL
22560-16-3
LiB(C2H5)3H
105.94
0.89
−17°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lithium triethylborohydride, 1.7M solution in THF

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Documents