Learn More
Thermo Scientific™ LN2 Level Monitor
Safeguard irreplaceable biological specimens with the LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available.
1590.10 € - 2470.00 €
Product Code | Brand | Voltage | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | For Use With (Equipment) | Price | Quantity & Availability | ||||
10164623
|
Thermo Scientific™
CN509X7 |
230 V | Locator 4 Plus |
2420.00 €
Each |
In Stock
Log in to see stock. |
||||
10052733
|
Thermo Scientific™
CN509X7-70 |
100/240 V | Locator 8 Plus |
2470.00 €
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
10174623
|
Thermo Scientific™
CN509X1-70 |
100/240 V | Locator Jr. Plus |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Safeguard irreplaceable biological specimens with the Thermo Scientific™ LN2 Level Monitor, one of the most advanced liquid nitrogen level monitors available. Designed to minimize liquid nitrogen evaporation, the monitor is for use with Thermo Scientific™ Locator™ and Locator Plus cryogenic storage systems.
Ultrasonic Level Monitor
- Minimizes conduction and liquid nitrogen evaporation
- Easy-to-read indicator continuously displays liquid nitrogen volume in 1/8 increments
- Audible and visual alarms signal low level liquid nitrogen conditions
- Warranty varies by country, please contact us for details
Specifications
LN2 Measuring Device | |
Alarm |