Learn More
LogIT Microsense Sensors
Especially designed to work with the DataMeter, LIVE, and LogIT family of products.
Brand: DCP Microdevelopments CRD-130-555T
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Identify themselves to datalogger as soon as they are plugged in
- Automatically calibrate and make the most effective use of the analogue or digital information
- Most are self-contained within a metal housing which encloses and protects the circuitry inside
- Remote leads available for more flexible experiment set-up
- Designed specifically for use in schools
- Strong Din-type connections which have proven their reliability over many years
Specifications
Sensor, Microsense | |
Most LogIT Dataloggers | |
0 to 200 beats/min. |
Measures Heart Rate | |
±10% |