missing translation for 'onlineSavingsMsg'
Learn More

Acros Organics™ Lopinavir, 98%

Brand:  Acros Organics™ 456750050

Code : NEW

Additional Details : CAS Number : 192725-17-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15325088

  • 391.70 € / 5g

EU Stock 2

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

192725-17-0
628.814
133109001
CC1=C(C(=CC=C1)C)OCC(=O)NC(CC2=CC=CC=C2)C(CC(CC3=CC=CC=C3)NC(=O)C(C(C)C)N4CCCNC4=O)O
C37H48N4O5
KJHKTHWMRKYKJE-PYJUJTJGSA-N
N-[(2R,4R,5R)-5-[[2-(2,6-dimethylphenoxy)acetyl]amino]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamide

Specifications

5g
C37H48N4O5
CC1=C(C(=CC=C1)C)OCC(=O)NC(CC2=CC=CC=C2)C(CC(CC3=CC=CC=C3)NC(=O)C(C(C)C)N4CCCNC4=O)O
628.814
192725-17-0
KJHKTHWMRKYKJE-PYJUJTJGSA-N
N-[(2R,4R,5R)-5-[[2-(2,6-dimethylphenoxy)acetyl]amino]-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-(2-oxo-1,3-diazinan-1-yl)butanamide
133109001
Videos
Documents