Learn More
Lovibond™ Glycine Tablet
Lovibond Glycine Tablet, Range: 0.01 to 3.8mg/L, White, Composition: Non-hazardous components 100%, Format: Tablet
Brand: Lovibond™ 512170BT
Code : NEW
Additional Details : CAS Number : 10043-35-3 Weight : 0.00100kg
Description
Lovibond Glycine Tablet, Range: 0.01 to 3.8mg/L, White, Composition: Non-hazardous components 100%, Format: Tablet
Specifications
White | |
5.8 | |
Water analysis, chlorine dioxide aqua comparator™ test kit, ozone aqua comparator™ test kit, chlorine dioxide reagent set, ozone reagent set | |
0.01 to 3.8mg/L | |
DPD |
Non-hazardous components 100% | |
Tablet | |
MC500 colorimeter, SP600 spectrophotometer, SC450 colorimeter | |
20°C | |
Glycine Tablet |