Learn More
Lovibond™ Sulfate 4 Powder Packs™
Premeasured unit dose reagent for water quality testing
Brand: Lovibond™ 532160
Additional Details : CAS Number : 10326-27-9 Weight : 0.09200kg Transport : UN number : 1564 Chem class : 6.1 Pack group : III
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
US EPA | |
Citric Acid 50 to 60%, Barium Chloride Dihydrate 40 to 50% | |
10 mL | |
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing | |
100 Pack | |
20°C | |
0.33 oz. |
White | |
2.3 | |
Powder | |
Hach™ and other manufacturers test kits, colorimeters and spectrophotometers | |
0 to 70mg/L | |
Sulfate 4 Reagent |
Safety and Handling
- vario sulphate 4
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : citric acid, bariumchloride dihydrate