missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Copper No. 1 Tablets
Brand: Lovibond™ 513550BT
Code : NEW
Additional Details : Weight : 0.30000kg
Specifications
Yellow | |
6 | |
Water analysis | |
0.05 to 5mg/L | |
Copper No. 1 Tablet |
30 to 40% Boric acid, 30 to 40% Potassium Dihydrogenorthophosphate, 1 to 10% Disodium tetraborate, anhydrous, 2.5% 2,2'-Bichinolin-4,4' Dicarbon acid Dipotassium Trihydrate p.a. 95%, 25 to 30% Non-hazardous components | |
Tablet | |
Colorimeter and spectrophotometer | |
Biquinoline |