missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: DEHA Tablets
Brand: Lovibond™ 513220BT
Code : NEW
Additional Details : Weight : 0.30000kg
Specifications
Yellow | |
5.6 | |
Water analysis | |
20 to 500μg/L | |
PPST |
Boric Acid 5 to 10%, non-hazardous components 90 to 100% | |
Tablet | |
Colorimeter and spectrophotometer | |
20°C | |
DEHA Tablet |