Learn More
Thermo Scientific Chemicals Low Form Inconel Crucible
79.90 € - 295.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
15423585
|
Thermo Scientific™
035886.KT |
15 mL | 33mm | 23mm |
90.80 €
Each |
In Stock
Log in to see stock. |
|||
15453595
|
Thermo Scientific™
035928.KT |
25 mL | 45mm | 23mm |
94.90 €
Each |
In Stock
Log in to see stock. |
|||
15473605
|
Thermo Scientific™
035970.KT |
55 mL | 46mm | 35mm |
111.00 €
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
15413615
|
Thermo Scientific™
035994.KT |
55 mL | 47mm | 43mm |
86.30 €
Each |
In Stock
Log in to see stock. |
|||
15473615
|
Thermo Scientific Chemicals
036016.KT |
75 mL | 51mm | 43mm |
79.90 €
Each |
In Stock
Log in to see stock. |
|||
15453625
|
Thermo Scientific Chemicals
036037.KT |
500 mL | 102mm | 66mm |
295.00 €
Each |
In Stock
Log in to see stock. |
|||
15403635
|
Thermo Scientific Chemicals
036056.KT |
100 mL | 59mm | 46mm |
166.00 €
Each |
In Stock
Log in to see stock. |
|||
15453635
|
Thermo Scientific™
036085.KT |
250 mL | 82mm | 60mm |
194.00 €
Each |
In Stock
Log in to see stock. |
|||