missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcelaines Avignon™ Low Form Porcelain Crucibles
Heat and cool substances in the laboratory using Porcelaines Avignon™ Low Form Porcelain Crucibles. They are available in various capacities.
Brand: Porcelaines Avignon™ 11306
Code : A0
Additional Details : Weight : 0.19260kg
Description
- Low walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifications
Porcelain | |
40 mL | |
36mm |
Glazed | |
55mm | |
Crucible |