Learn More
Thermo Scientific Chemicals Low Form Zirconium Crucible
93.15 € - 281.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Height (Metric) | Price | Quantity & Availability | |||
15473585
|
Thermo Scientific Chemicals
035905.KT |
20 mL | 33mm | 30mm |
141.00 €
Each |
In Stock
Log in to see stock. |
|||
15433595
|
Thermo Scientific Chemicals
035926.KT |
25 mL | 45mm | 23mm |
93.20 €
Each |
In Stock
Log in to see stock. |
|||
15493595
|
Thermo Scientific Chemicals
035947.KT |
35 mL | 46mm | 30mm |
164.00 €
Each |
In Stock
Log in to see stock. |
|||
15453605
|
Thermo Scientific Chemicals
035968.KT |
55 mL | 46mm | 35mm |
104.00 €
Each |
In Stock
Log in to see stock. |
|||
15433615
|
Thermo Scientific Chemicals
035997.KT |
55 mL | 47mm | 43mm |
192.00 €
Each |
In Stock
Log in to see stock. |
|||
15493615
|
Thermo Scientific™
036018.KT |
75 mL | 51mm | 43mm |
125.00 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
15473625
|
Thermo Scientific™
036039.KT |
500 mL | 102mm | 66mm |
281.00 €
Each |
In Stock
Log in to see stock. |
|||
15423635
|
Thermo Scientific Chemicals
036058.KT |
250 mL | 82mm | 60mm |
178.00 €
Each |
In Stock
Log in to see stock. |
|||
15403585
|
Thermo Scientific™
035884.KT |
15 mL | 33mm | 23mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||