missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAM-1 Rat, Biotin, Clone: DATK32, enQuireBio™
Rat Monoclonal Antibody
82.15 € - 143.00 €
Specifications
Antigen | LPAM-1 |
---|---|
Clone | DATK32 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932418
|
EnquireBio
QAB73-B-25UG |
25 μg |
82.15 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922418
|
EnquireBio
QAB73-B-100UG |
100 μg |
143.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LPAM-1 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Primary | |
Store at 2-4°C. |
DATK32 | |
Flow Cytometry | |
Biotin | |
RUO | |
IgG2a κ | |
Protein A/G Purified |