missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSECtin/CLEC4G Antibody (845404), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2947D
533.80 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
LSECtin/CLEC4G Monoclonal antibody specifically detects LSECtin/CLEC4G in Human samples. It is validated for Flow Cytometry
Specifications
| LSECtin/CLEC4G | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 339390 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 845404 | |
| Flow Cytometry | |
| C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, UNQ431 | |
| NS0 mouse myeloma cell line transfected with LSECtin/CLEC4G, Ser54-Cys293, Accession # Q6UXB4 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction