missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ [Lys&superscript_1;, Pro&superscript_2;,&superscript_5;, Arg&superscript_3;,&superscript_4;, Tyr&superscript_6;] Vasoactive Intestinal Peptide, Human, Porcine, Rat

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66794.|D

Code : NEW

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15415815

  • 128.00 € / 0.50mg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Specifications

3467.10
Human, Porcine, Rat,
Solid
RUO
[Lys1, Pro25, Arg34, Tyr6] VIP, human, porcine, rat
HPLC
0.5mg
Lys-Pro-Arg-Arg-Pro-Tyr-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2
-30° to -10°C
[Lys1, Pro2,5, Arg3,4, Tyr6] Vasoactive Intestinal Peptide, human, porcine, rat
Videos
SDS

For Research Use Only