missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MabSelect Sure™ LX Chromatography Media
Protein A affinity medium with high dynamic binding capacity which improves performance and cuts costs
4007.00 € - 8713.00 €
Specifications
For Use With (Application) | For PurifiCatIon of Monoclonal Antibodies for Clinical AppliCatIons |
---|---|
Stability | Stable in aqueous Buffers commonly used in Protein A Chromatography |
Storage Requirements | 2°C to 8°C |
Binding Capacity | 60 mg Human IgG/mL mediµm |
Product Type | MabSelect SuRe LX Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11683074
|
Cytiva
17-5474-01 |
25 mL |
4007.00 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
11693074
|
Cytiva
17-5474-02 |
200 mL |
8713.00 €
200mL |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
Alkali tolerance allows for stability, thorough cleaning and reduction in contamination
- Alkali-stabilized Protein A-derived (E.coli) Ligand
- High dynamic binding capacity (DBC) for high titer cultures
- Reduces risk of contamination and carryover
- Alkali-tolerant rProtein A ligand withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
Purification of monoclonal antibodies for clinical applications and process scale
Certifications
Certificate of Analysis
Specifications
For PurifiCatIon of Monoclonal Antibodies for Clinical AppliCatIons | |
2°C to 8°C | |
MabSelect SuRe LX Media | |
85 μm | |
20 cm | |
<2 bar | |
3 to 12 | |
Alkali-stabilized Protein A-derived (E. coli) |
Stable in aqueous Buffers commonly used in Protein A Chromatography | |
60 mg Human IgG/mL mediµm | |
500 cm/Hoµr | |
7.87 in. | |
Certificate of Analysis | |
Highly Cross-Linked Agarose | |
20% Ethanol |