Learn More
Magnesium bromide, 98%, pure, anhydrous, ACROS Organics™
Brand: Acros Organics 413390050
Code : 29
Additional Details : CAS Number : 7789-48-2 Weight : 0.05500kg
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
7789-48-2 | |
184.11 | |
OTCKOJUMXQWKQG-UHFFFAOYSA-L | |
82241 | |
[Mg+2].[Br-].[Br-] |
Br2Mg | |
MFCD00011105 | |
magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent | |
magnesium;dibromide |
Specifications
Magnesium bromide | |
7789-48-2 | |
Br2Mg | |
MFCD00011105 | |
15,5723 | |
Solubility in water: 1015g/L (20°C). Other solubilities: slightly soluble in alcohol | |
[Mg+2].[Br-].[Br-] | |
184.11 | |
184.11 | |
Pure | |
Solubility in water: 1015g/L (20°C). Other solubilities: slightly soluble in alcohol | |
5g |
Anhydrous | |
97.5 to 102.5% (Argentometry) | |
MgBr2 | |
02,254; 06,270; 08,273; 10,14; 15,194; 16,199 | |
magnesium bromide, magnesium dibromide, dibromomagnesium, magnesium-bromide, acmc-20aklq, magnesium bromide salt, magnesium ii bromide, magnesium 2+ ion dibromide, grignard reagent | |
OTCKOJUMXQWKQG-UHFFFAOYSA-L | |
magnesium;dibromide | |
82241 | |
98% | |
Glass bottle | |
711.0°C |
Safety and Handling
- Magnesium bromide, anhydrous
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.