missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, purum p.a, ≥98.0% (KT), Honeywell™
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Melting Point | 117°C |
Color | Colorless |
pH | 5.0 to 6.0 (at 20°C) |
Physical Form | Crystalline |
Quantity | 5 kg |
Linear Formula | MgCl2 · 6H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 203.3g/mol |
Percent Purity | ≥98.0% (KT) |
Packaging | Poly bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'