missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, Extra Pure, Light, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Brand:  Fisher Chemical M/0850/60

Code : 21

Additional Details : CAS Number : 1309-48-4 Weight : 1.00000kg

Packaging Plastic keg
Quantity 1kg

 View more versions of this product

Product Code. 10040240

  • 114.18 € / 1kg
Estimated Shipment: 30-05-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1309-48-4
40.304
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
11109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium

Specifications

1309-48-4
White
10.3
MgO
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
O=[Mg]
40.304
40.29g/mol
Plastic keg
3600°C
2800°C
1kg
11109
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
Powder Solid
0 hPa at 20°C
Videos
SDS Documents
Certificates
Special Offers

Special Offers