missing translation for 'onlineSavingsMsg'
Learn More

Magnesium stearate, Thermo Scientific Chemicals

Catalog Number p-7122667
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
557-04-0
C36H70MgO4
591.257
MFCD00036391
HQKMJHAJHXVSDF-UHFFFAOYSA-L
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
11177
CHEBI:9254
magnesium;octadecanoate
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]
This item is not returnable. View return policy
557-04-0
C36H70MgO4
591.257
MFCD00036391
HQKMJHAJHXVSDF-UHFFFAOYSA-L
magnesium stearate, magnesium octadecanoate, magnesium distearate, synpro 90, octadecanoic acid, magnesium salt, dibasic magnesium stearate, petrac mg 20nf, ns-m salt, sm-p, stearic acid, magnesium salt
11177
CHEBI:9254
magnesium;octadecanoate
CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Mg+2]

Magnesium stearate is involved in the production of medical tablets, capsules, powders and polymer formulation. It is also used as a lubricant for tablets, anti-adherent, in dry coating and as a binding agent. It is an important ingredient in baby formulas. Further, it is used in the hydrogenation process. In addition to this, it is used to bind the sugar in hard candies like mints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in benzene. Insoluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, acids and alkalis.
TRUSTED_SUSTAINABILITY
Melting Point 148°C to 152°C
Quantity 250 g
Solubility Information Slightly soluble in benzene. Insoluble in water,alcohol and ether.
Formula Weight 591.27
Chemical Name or Material Magnesium stearate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'