missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Magnetic beadsTM Streptavidin
Click to view available options
Quantity:
5 mg
10 mg
Packungsgröße:
10mg
5mg
Beschreibung
The streptavidin magnetic beads are uniform size, narrow size distribution with large surface area and unique surface coating, it is easy to capture the biotinylated proteins or other molecules, and the bounded protein have no activity lost, can help us get the best performance and highly reproducible results.
Spezifikation
Spezifikation
| Kit Contents | Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L, the Biotinylated protein can bind to the SA beads irreversibly. The Magenetic beads are pre-coupled with Streptavidin tetramer |
| Content And Storage | -20°C, Avoid light |
| Shipping Condition | RT |
| Quantity | 10 mg |
| Type | Magnetic Affinity Bead |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur