missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Magnetic Filters Funnel with Lid

Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes.

Brand:  Cytiva (formerly Pall Lab) 4241

Additional Details : Weight : 0.32300kg

Product Code. 17134281

  • 408.00 € / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

47 mm
Polyphenylsulfone Cylinder and Stem, Silicone O-Ring, Polypropylene Vent Plug
Non-sterile
300 mL
Lid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers