missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Magnetic Filters Funnel Top Only

Replacement housing for funnels with unique magnetic seal.

Brand:  Cytiva (formerly Pall Lab) 4254

Additional Details : Weight : 0.00100kg

Product Code. 17134291

  • 285.00 € / Each
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

Polyphenylsulfone
Top Only
47 mm
Non-sterile
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers