missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Chloride Tetrahydrate, Extra Pure, SLR, Fisher Chemical

Brand:  Fisher Chemical M/1800/53

Code : 21

Additional Details : CAS Number : 13446-34-9 Weight : 0.50000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 500g

 View more versions of this product

Product Code. 10010260

Quantity Price
1 68.23 € / 500g
4 63.76 € / 500g
Estimated Shipment
06-08-2020

Please to purchase this item. Need a web account? Register with us today!



You may only order up to the available stock. For more details call customer services on 01 885 5854 .

Specifications

Chemical Identifiers

13446-34-9
Cl2Mn·4H2O
149792

Specifications

13446-34-9
149792
Solid Powder
Red
4 to 6
Cl2Mn·4H2O
197.91g/mol
Plastic powder jar
58°C
500g
Videos
SDS Documents