Learn More
Socorex™ Acura Manual 810 Dilution Micropipettes
Use graduated glass pipettes when performing serial 1:10 dilutions in bacteriology with Socorex™ Acura Manual 810 Dilution Micropipettes. They feature smooth activation with excellent ergonomics.
Brand: Socorex™ 810.1100
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Three-year warranty
- No pre-calibrated fixed volumes
- No setting required
- Independent calibration for each volume
- Autoclavable
Specifications
CE Marked, IVD | |
Manual 810 Dilution | |
Dilution Pipette | |
1 Each |
Autoclavable | |
Acura™ | |
1 +0.1 mL |