missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCO Antibody (2359A), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB29561MFV450
510.00 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| MARCO | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 8685 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 2359A | |
| Flow Cytometry, CyTOF-ready | |
| macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2 | |
| Mouse myeloma cell line, NS0-derived mouse MARCO protein, Gln70-Ser518, Accession # Q60754 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction