missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 0.5mL ScrewTop Tubes in Barcoded Latch Racks

Ultimate sample traceability at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 0.5mL ScrewTop Tubes in barcoded latch racks.

Brand:  Thermo Scientific™ 3744-BR

379.14 EUR valid until 2019-02-28
Use promo code "13474" to get your promotional price.



Code : NEW

Additional Details : Weight : 1.15000kg

 View more versions of this product

Product Code. 15400004

  • 471.51 € / Pack of 480



UK Stock 19
Add to basket

Specifications

Specifications

Virgin Class VI Medical Grade Polypropylene with Virgin Class VI Medical Grade Silicone O-ring
Yes
Colorless
0.5mL
Virgin Class VI Medical Grade Polypropylene
0.5mL
ScrewTop, 2D Barcode
Sterile
Clear
V-Bottom ScrewTop Tubes
Yes, with cap loosened
V-Bottom
5 racks of 96/case
Tube barcode: Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction; Rack barcode: 2D Datamatrix; Linear EAN Code 128; Human Readable
Matrix™
Videos