Learn More
MDM2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27883-1-AP-20UL
This item is not returnable.
View return policy
Description
The MDM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MDM2. This antibody recognizes human antigen. The MDM2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| MDM2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 4193 | |
| IgG |
| Western Blot, Flow Cytometry | |
| 367 μg/mL | |
| Rabbit | |
| Primary | |
| Human |