Learn More
Hirschmann™ Class B Volumetric Flask with Stopper
Calibrated to contain a precise volume at a specific temperature. Flasks are used for precise dilutions and prepare standard solutions.
62.70 € - 91.75 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10627687
|
Hirschmann™
7550181 |
100 mL |
90.30 €
Pack of 6 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
13128059
|
Hirschmann™
7550175 |
50 mL |
83.40 €
Pack of 6 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
13118059
|
Hirschmann™
7550170 |
25 mL |
66.00 €
Pack of 6 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
13108059
|
Hirschmann™
7550160 |
10 mL |
62.70 €
Pack of 6 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
13148059
|
Hirschmann™
7550186 |
250 mL |
91.75 €
Pack of 5 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
13158059
|
Hirschmann™
7550190 |
500 mL |
77.40 €
Pack of 4 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
10608962
|
Hirschmann™
7550193 |
1000 mL |
66.30 €
Pack of 3 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN EN ISO 1042
- PP
- Adjusted ringmark
- translucent
- with standard ground poly stoppers