missing translation for 'onlineSavingsMsg'
Learn More

Manutan Medium duty tubular starter shelving, blue, 2000mm x 1250mm x 500mm

Brand:  Manutan A360386

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12857998

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Economic, multi-purpose shelving with easy 'click-in' system. Ideal for various applications from a simple archive shelving to a complete warehousing operation. Perforated every 62. 5mm for shelf adjustment. Simple assembly - simple 'click-in' shelves. Tubular shelves are equipped with welded hooks for easy connection to uprights with all components powder coated for extra durability.
  • Five shelves
  • Blue uprights
  • Shelf capacity UDL: 132kg
  • Dimensions (H x W x D): 2000mm x 1250mm x 500mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers