Learn More
Emma Safety Footwear Mendoza Safety Shoes
State-of-the art Sympatex™ construction provides perfect foot protection for hands-on roles within the Chemical Industry and Agriculture. Virtually waterproof, the Mendoza boots allow feet to breathe properly, keeping them cool and dry.
Brand:  Emma Safety Footwear MM58284842
Additional Details : Weight : 2.22300kg
 View more versions of this product
Product Code. 15958781
  • 214.12 € / 1 pair
Estimated Shipment: 26-09-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck / Sympatex™ construction
  • Waterproof
  • GripForce™ Duo Heat Resistant Outsole
Specifications
Specifications
EN ISO 20345:2011
Mendoza
Oiled Nubuck
Steel
Brown
Hydro-Tec™ Sanitized Silver / Sympatex™
42