missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mercury(I) nitrate dihydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Beschreibung
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
Spezifikation
Melting Point | 70°C |
Quantity | 50 g |
Linear Formula | Hg2(NO3)2·2H2O |
UN Number | UN1627 |
Merck Index | 14,5896 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 561.22 (525.19 Anhydrous) |
Percent Purity | 98% |
Odor | Odorless |
Sensitivity | Light sensitive |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'