missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS-CoV Spike Protein Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18800MFV500
492.15 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
MERS-CoV Spike Protein Polyclonal antibody specifically detects MERS-CoV Spike Protein in MERS-CoV samples. It is validated for Western Blot, ELISA
Specifications
| MERS-CoV Spike Protein | |
| Polyclonal | |
| Western Blot, ELISA | |
| MERS S glycoprotein, MERS S1, MERS S2, MERS Spike glycoprotein, MERS Spike Protein, MERS-CoV S glycoprotein, Middle East respiratory syndrome-related coronavirus Spike glycoprotein, Spike glycoprotein, spike protein, Spike RBD, surface glycoprotein | |
| Produced in rabbits immunized with purified, recombinant MERS-CoV Spike Protein (Accession#: AFS88936.1; Asp726-Pro1296). | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 14254594 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| MERS-CoV | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction