missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitroso-dimethylamine in Methanol 10μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Specifications
Specifications
| Packaging | Ampoule(s) |
| CAS | 62-75-9 |
| Molecular Formula | C2H6N2O |
| Concentration | 10mu;g/mL |
| UN Number | 1230 |
| InChI Key | UMFJAHHVKNCGLG-UHFFFAOYSA-N |
| SMILES | CN(C)N=O |
| IUPAC Name | N,N-dimethylnitrous amide |
| Solvent or Matrix | Methanol |
| Molecular Weight (g/mol) | 74.083 |
| Show More |
Safety and Handling
Product Identifier
- Standard solution Dimethylnitrosoamine,N-nitrosodimethylamine 10ug/ml in methanol
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction