missing translation for 'onlineSavingsMsg'
Learn More

Methanol, AMD CHROMASOLV™, ≥99.9%, Solstice

CHROMASOLV™ AMD,≥99.9%

72.05 € - 250.80 €


Products 2
Product Code Brand dimQuantity Packaging Price Quantity & Availability  
Product Code Brand dimQuantity Packaging Price Quantity & Availability  
15694690
SDS View Documents Promotion Details
Honeywell-Chromasolv
34898-2.5L
ONLINE EXCLUSIVE PRICE
88.00 €
72.05 € / 2.5 L
Save 15.95 €
18% Off
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15604700
SDS View Documents Promotion Details
Honeywell-Chromasolv
34898-4X2.5L
ONLINE EXCLUSIVE PRICE
306.00 €
250.80 € / Pack of 4
Save 55.20 €
18% Off
View alternative products
Save Up to 
»
N/A
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description

Description

  • Solvents designed for high-performance thin-layer chromatography
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: methanol

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.