missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED HPLC, J.T. Baker™

Product Code. p-7138043 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
Unit Size:
1 L
2.5 L
5 L
This item is not returnable. View return policy

Product Code. 10241061

Brand: JT Baker 8404.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Get rapid, reproducible performance and separation results. Methanol, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use

Specifications

Melting Point -97.8°C
Absorbance Max. 0.30 at 225nm, Max. 0.10 at 240nm, Max. 0.01 at 265nm
Color Colorless
CAS 67-56-1
Density 0.79 g/mL at 25°C
Boiling Point 64°C
Quantity 1 L
Explosion Limit 6 - 36 Vol %
Flash Point 11°C
Ignition Point 240°C
Odor Alcohol
Packaging Glass
Physical Form Liquid
Refractive Index 1.3314 nD
Residue after Evaporation Max. 5 ppm
Molecular Formula CH4O
Titratable Acid Max. 0.0003meq/g
Linear Formula CH3OH
Titratable Base Max. 0.0002meq/g
Vapor Pressure 16.9 kPa at 25°C
Water Max. 0.05%
Synonym Carbinol, Methyl alcohol, Wood alcohol
Formula Weight 32.04g/mol
Percent Purity ≥99.8%
Grade HPLC Isocratic Grade
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.